Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749821.3 | XP_003749869.1 | 6 - 7 | 1088 | 72 | |
XM_008763272.1 | XP_008761494.1 | 6 - 7 | 1088 | 72 | ||
XM_575654.6 | XP_575654.3 | 6 - 7 | 1088 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |