Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107890.1 | NP_001101360.1 | 4 - 5 | 801 | 86 | |
XM_006237494.2 | XP_006237556.1 | 8 - 9 | 932 | 86 | ||
XM_006237495.2 | XP_006237557.1 | 7 - 8 | 748 | 86 | ||
XM_006237496.2 | XP_006237558.1 | 7 - 8 | 811 | 86 | ||
XM_006237498.2 | XP_006237560.1 | 6 - 7 | 631 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |