Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247907.2 | XP_006247969.1 | 1 - 2 | 752 | 63 | |
GenBank mRNA | FQ212922.1 | - | 1 - 2 | 220 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |