Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113781.2 | NP_001107253.2 | 5 - 6 | 762 | 90 | |
XM_006246759.2 | XP_006246821.1 | 5 - 6 | 872 | 90 | ||
GenBank mRNA | BC158692.1 | - | 5 - 6 | 762 | 9022735 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |