Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc42b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_222198.7 | XP_222198.5 | 3 - 4 | 486 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |