Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107130.2 | NP_001100600.2 | 9 - 10 | 1482 | 123 | |
XM_006249139.2 | XP_006249201.1 | 9 - 10 | 1995 | 123 | ||
XM_008769135.1 | XP_008767357.1 | 9 - 10 | 2064 | 123 | ||
XM_008769136.1 | XP_008767358.1 | 10 - 11 | 2086 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |