Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106586.1 | NP_001100056.1 | 4 - 5 | 537 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |