Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107855.1 | NP_001101325.1 | 6 - 7 | 2071 | 107 | |
XM_006236326.2 | XP_006236388.1 | 5 - 6 | 1947 | 107 | ||
GenBank mRNA | BC161918.1 | - | 5 - 6 | 1859 | 8369971 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |