Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108516.1 | NP_001101986.1 | 4 - 5 | 514 | 67 | |
XM_006230850.1 | XP_006230912.1 | 4 - 5 | 539 | 67 | ||
GenBank mRNA | BC166788.1 | - | 4 - 5 | 461 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |