Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012887.1 | NP_037019.1 | 7 - 8 | 1233 | 111 | |
XM_008765219.1 | XP_008763441.1 | 5 - 6 | 1056 | 111 | ||
GenBank mRNA | U18314.1 | - | 7 - 8 | 1233 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |