Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008378.1 | NP_001008379.1 | 4 - 5 | 538 | 62 | |
GenBank mRNA | BC086452.1 | - | 4 - 5 | 538 | 7307024 | 62 |
FQ214182.1 | - | 4 - 5 | 508 | 62 | ||
FQ218380.1 | - | 4 - 5 | 508 | 62 | ||
FQ219469.1 | - | 4 - 5 | 531 | 62 | ||
FQ219661.1 | - | 4 - 5 | 536 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |