Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012121.1 | NP_001012121.1 | 3 - 4 | 445 | 106 | |
XM_008765722.1 | XP_008763944.1 | 4 - 5 | 1541 | 106 | ||
XM_008765723.1 | XP_008763945.1 | 3 - 4 | 301 | 106 | ||
XM_008765724.1 | XP_008763946.1 | 3 - 4 | 1126 | 106 | ||
XM_008765725.1 | XP_008763947.1 | 4 - 5 | 278 | 106 | ||
XM_008765726.1 | XP_008763948.1 | 4 - 5 | 273 | 106 | ||
GenBank mRNA | BC090007.1 | - | 3 - 4 | 445 | 7323849 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |