Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106285.1 | NP_001099755.1 | 1 - 2 | 254 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |