Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135858.1 | NP_001129330.1 | 9 - 10 | 1481 | 112 | |
XM_008765316.1 | XP_008763538.1 | 8 - 9 | 1391 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |