Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241426.2 | XP_006241488.1 | 4 - 5 | 2019 | 80 | |
XM_006241428.2 | XP_006241490.1 | 3 - 4 | 1772 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |