Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130411.2 | NP_569095.1 | 9 - 10 | 1175 | 72 | |
XM_006230204.2 | XP_006230266.1 | 9 - 10 | 1259 | 72 | ||
GenBank mRNA | AF416730.1 | - | 8 - 9 | 1071 | 72 | |
AF495469.1 | - | 9 - 10 | 1175 | 72 | ||
BC086971.1 | - | 9 - 10 | 1151 | 7124429 | 72 | |
FQ225277.1 | - | 6 - 7 | 697 | 72 | ||
FQ225360.1 | - | 6 - 7 | 697 | 72 | ||
FQ225522.1 | - | 5 - 6 | 614 | 72 | ||
FQ226290.1 | - | 7 - 8 | 644 | 72 | ||
FQ233306.1 | - | 9 - 10 | 1169 | 72 | ||
FQ233390.1 | - | 9 - 10 | 1171 | 72 | ||
FQ233414.1 | - | 9 - 10 | 1173 | 72 | ||
FQ233980.1 | - | 9 - 10 | 1089 | 72 | ||
FQ234750.1 | - | 9 - 10 | 1177 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |