Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191840.1 | NP_001178769.1 | 9 - 10 | 1001 | 158 | |
XM_006230349.2 | XP_006230411.1 | 9 - 10 | 1851 | 158 | ||
XM_006230350.2 | XP_006230412.1 | 9 - 10 | 973 | 158 | ||
XM_008759920.1 | XP_008758142.1 | 8 - 9 | 2058 | 158 | ||
XM_008759921.1 | XP_008758143.1 | 9 - 10 | 1058 | 158 | ||
GenBank mRNA | AF314960.1 | - | 5 - 6 | 581 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |