Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108204.1 | NP_001101674.1 | 2 - 3 | 444 | 75 | |
XM_006244537.1 | XP_006244599.1 | 1 - 2 | 87 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |