Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100709.1 | NP_001094179.1 | 8 - 9 | 970 | 71 | |
XM_006238932.2 | XP_006238994.1 | 9 - 10 | 1137 | 71 | ||
XM_006238933.2 | XP_006238995.1 | 9 - 10 | 1099 | 71 | ||
XM_008764140.1 | XP_008762362.1 | 9 - 10 | 1155 | 71 | ||
GenBank mRNA | BC099218.1 | - | 4 - 5 | 574 | 71 | |
BC160881.1 | - | 8 - 9 | 970 | 7381544 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |