Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108676.1 | NP_001102146.1 | 9 - 10 | 1170 | 116 | |
XM_006238804.2 | XP_006238866.1 | 9 - 10 | 1715 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |