Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240356.2 | XP_006240418.1 | 1 - 2 | 503 | 87 | |
XM_006240358.2 | XP_006240420.1 | 1 - 2 | 329 | 87 | ||
XM_008764889.1 | XP_008763111.1 | 1 - 2 | 326 | 87 | ||
XM_008764890.1 | XP_008763112.1 | 1 - 2 | 319 | 87 | ||
XM_343089.7 | XP_343090.4 | 1 - 2 | 315 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |