Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011984.1 | NP_001011984.1 | 4 - 5 | 657 | 61 | |
XM_006240464.2 | XP_006240526.1 | 2 - 3 | 1302 | 61 | ||
GenBank mRNA | BC085882.1 | - | 4 - 5 | 657 | 7191983 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |