Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106697.1 | NP_001100167.1 | 6 - 7 | 925 | 61 | |
XM_006239178.2 | XP_006239240.1 | 6 - 7 | 793 | 61 | ||
XM_008764230.1 | XP_008762452.1 | 5 - 6 | 527 | 61 | ||
GenBank mRNA | BC168898.1 | - | 6 - 7 | 492 | 61 | |
FQ227354.1 | - | 6 - 7 | 538 | 61 | ||
FQ227472.1 | - | 6 - 7 | 777 | 61 | ||
FQ234412.1 | - | 6 - 7 | 777 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |