Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004090.2 | NP_001004090.2 | 4 - 5 | 454 | 103 | |
GenBank mRNA | AF540877.1 | - | 4 - 5 | 480 | 103 | |
BC100135.1 | - | 4 - 5 | 454 | 7315329 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |