Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024358.1 | NP_001019529.2 | 2 - 3 | 206 | 101 | |
GenBank mRNA | BC083665.1 | - | 2 - 3 | 206 | 7135489 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |