Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009543.1 | NP_001009543.1 | 420 | 78 | ||
GenBank mRNA | BC060590.1 | - | 420 | 6918453 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |