Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024368.1 | NP_001019539.1 | 2 - 3 | 110 | 85 | |
XM_006229606.2 | XP_006229668.1 | 2 - 3 | 867 | 85 | ||
XM_008773611.1 | XP_008771833.1 | 2 - 3 | 250 | 85 | ||
GenBank mRNA | BC086556.1 | - | 2 - 3 | 110 | 7304843 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |