Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271214.1 | NP_001258143.1 | 10 - 11 | 1388 | 81 | |
XM_008771657.1 | XP_008769879.1 | 10 - 11 | 1393 | 81 | ||
XM_008771658.1 | XP_008769880.1 | 11 - 12 | 1707 | 81 | ||
XM_008771659.1 | XP_008769881.1 | 11 - 12 | 1777 | 81 | ||
XM_008771660.1 | XP_008769882.1 | 9 - 10 | 1159 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |