Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 9 - 10 | 824 | 72 | |
XM_006257329.2 | XP_006257391.1 | 9 - 10 | 896 | 72 | ||
XM_006257330.2 | XP_006257392.1 | 9 - 10 | 1305 | 72 | ||
GenBank mRNA | M88751.1 | - | 9 - 10 | 828 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |