Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013200.1 | NP_001013218.1 | 5 - 6 | 920 | 152 | |
GenBank mRNA | BC083744.1 | - | 5 - 6 | 920 | 7192147 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |