Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138509.3 | NP_612518.2 | 3 - 4 | 344 | 152 | |
XM_006235294.2 | XP_006235356.1 | 3 - 4 | 327 | 152 | ||
XM_006235295.2 | XP_006235357.1 | 3 - 4 | 397 | 152 | ||
GenBank mRNA | BC081726.1 | - | 3 - 4 | 344 | 7192469 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |