Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106525.3 | NP_001099995.1 | 2 - 3 | 278 | 84 | |
GenBank mRNA | BC168917.1 | - | 2 - 3 | 278 | 7132786 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |