Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255055.2 | XP_006255117.1 | 1 - 2 | 187 | 69 | |
GenBank mRNA | FM054321.1 | - | 1 - 2 | 158 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |