Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031572.1 | NP_113760.1 | 6 - 7 | 966 | 115 | |
GenBank mRNA | BC089790.1 | - | 6 - 7 | 974 | 7371667 | 115 |
FQ209627.1 | - | 3 - 4 | 405 | 115 | ||
FQ219491.1 | - | 5 - 6 | 841 | 115 | ||
FQ219720.1 | - | 3 - 4 | 436 | 115 | ||
J03786.1 | - | 6 - 7 | 966 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |