Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181378.2 | NP_852043.1 | 6 - 7 | 836 | 112 | |
GenBank mRNA | AF456462.1 | - | 6 - 7 | 836 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |