Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766715.1 | XP_008764937.1 | 1 - 2 | 566 | 112 | |
XM_008766716.1 | XP_008764938.1 | 2 - 3 | 392 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |