Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243659.2 | XP_006243721.1 | 4 - 5 | 998 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |