Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191738.1 | NP_001178667.1 | 7 - 8 | 2665 | 68 | |
XM_006234311.1 | XP_006234373.1 | 7 - 8 | 1474 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |