Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177442.1 | NP_001170913.1 | 5 - 6 | 606 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |