Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107898.1 | NP_001101368.1 | 6 - 7 | 2409 | 66 | |
XM_006237743.2 | XP_006237805.1 | 7 - 8 | 2524 | 66 | ||
XM_008763495.1 | XP_008761717.1 | 7 - 8 | 2830 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |