Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107017.2 | NP_001100487.1 | 1 - 2 | 219 | 94 | |
GenBank mRNA | BC162047.1 | - | 1 - 2 | 219 | 7107394 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |