Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100568.1 | NP_001094038.1 | 1 - 2 | 148 | 84 | |
GenBank mRNA | CB718262.1 | - | 1 - 2 | 148 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |