Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025114.1 | NP_001020285.1 | 3 - 4 | 338 | 85 | |
XM_006256586.2 | XP_006256648.1 | 3 - 4 | 531 | 85 | ||
XM_006256587.2 | XP_006256649.1 | 3 - 4 | 532 | 85 | ||
GenBank mRNA | BC091322.1 | - | 3 - 4 | 338 | 7313558 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |