Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030027.1 | NP_001025198.1 | 6 - 7 | 642 | 92 | |
XM_006253313.2 | XP_006253375.1 | 6 - 7 | 665 | 92 | ||
GenBank mRNA | BC099741.1 | - | 6 - 7 | 642 | 5623828 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |