Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022851.1 | NP_074042.1 | 5 - 6 | 851 | 73 | |
GenBank mRNA | L34821.1 | - | 5 - 6 | 730 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |