Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107838.1 | NP_001101308.1 | 2 - 3 | 259 | 69 | |
GenBank mRNA | BC168896.1 | - | 2 - 3 | 239 | 7103021 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |