Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106562.1 | NP_001100032.1 | 7 - 8 | 1118 | 68 | |
XM_006233757.2 | XP_006233819.1 | 7 - 8 | 1109 | 68 | ||
XM_008761634.1 | XP_008759856.1 | 7 - 8 | 1025 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |