Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231152.2 | XP_006231214.2 | 5 - 6 | 7972 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |