Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252196.2 | XP_006252258.1 | 1 - 2 | 161 | 115 | |
XM_006252199.2 | XP_006252261.1 | 1 - 2 | 159 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |