Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767384.1 | XP_008765606.1 | 4 - 5 | 1206 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |